EMAIL THIS PAGE TO A FRIEND
Revista cubana de medicina tropical
[Determination in vivo of the role of esterase and glutathione transferase enzymes in pyrethroid resistance of Aedes aegypti (Diptera: Culicidae)].
PMID 23427458
Abstract
An in vivo study of two synergists, that is, Triphenil phosphate -specific esterase inhibitor- and ethacrynic acid -specific gluthation transferase inhibitor- was performed to determine if these enzymes were responsible for pyrethroid resistance of Aedes aegypti. To this end, two insecticide resistant Aedes aegypti strains were used, one strain selected with temephos by six selection generations (SAN-F6) and the other strain with delmamethrin by 12 selection generations (SAN-F12), being both strains resistant to pyrethroid insecticices. Through the use of TPP and EA synergists, it was proved that esterase and gluthation-s-transferase (GST) enzymes were responsible for pryrethroid resistance of these strains. These results showed the existence of cross-resistance and multidrug resistance, which should be taken into account for insecticide use strategies aimed at vector control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241288
Triphenyl phosphate, ≥99%
C18H15O4P
442829
Triphenyl phosphate, analytical standard
C18H15O4P