EMAIL THIS PAGE TO A FRIEND
Ugeskrift for laeger
[Dapsone-induced decrease in the HbA(1c) level may cause artefacts in diabetic control].
PMID 23428267
Abstract
Glycated haemoglobin (HbA(1c)) level is used to monitor glycaemic control in diabetic patients, and has recently been included as a diagnostic factor for type 2 diabetes mellitus (DM). A 50-year-old male with well-controlled type 1 DM, Mb Addison and coeliac disease was diagnosed with dermatitis herpetiformis and treated with dapsone. Subsequently, the patient presented a pronounced drop in HbA(1c) levels without changes in blood glucose levels. S-fructosamine values were in agreement with the previous stable HbA(1c) values. Blood analyses showed inappropriately low HbA(1c) values due to mild haemolysis and shortening of erythrocyte lifetime without anaemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A74807
4-Aminophenyl sulfone, 97%
C12H12N2O2S
46158
Dapsone, VETRANAL®, analytical standard
C12H12N2O2S