EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Discrimination of viable from non-viable gram-negative bacterial pathogens in airborne particles using propidium monoazide-assisted qPCR.
PMID 23428754
Abstract
The presence of bacterial pathogens in airborne particulate matter (PM) has been of considerable concern from the public health standpoint. Conventional culture-based methods are tedious, time consuming and are unable to quantify stressed viable but non-culturable (VBNC) populations of these pathogens. This study reports the optimization, validation and application of a new and rapid quantitative method for enumeration of four live potential Gram-negative bacterial pathogens (Escherichia coli, Klebsiella pneumoniae, Pseudomonas aeruginosa and Aeromonas hydrophila) in PM of biomass burning origin. This method makes use of an intercalating dye (propidium monoazide, PMA) in conjunction with real-time PCR (qPCR) analysis following DNA extraction from PM samples for distinguishing viable from non-viable potential bacterial pathogens. This method was not affected by the complex matrix of the environmental samples, nor by any PCR inhibition effects. The number of viable pathogens ranged from 0 to 8×10(4) gene copies/m(3) in PM. With the exception of A. hydrophilia, all the three pathogens were found to be present in PM. The correlation between the counts obtained using the PMA-qPCR (modified qPCR) and those from the culture-based method was very high with R(2)~1.0 and p value<0.0001.