EMAIL THIS PAGE TO A FRIEND
Asian cardiovascular & thoracic annals
Traumatic thoracobiliary (pleurobiliary and bronchobiliary) fistula.
PMID 23430419
Abstract
Traumatic thoracobiliary fistula is a rare but serious complication. A series of thoracobiliary fistulas secondary to penetrating trauma and analysis of trends in management are presented. We retrospectively reviewed all patients with traumatic thoracobiliary fistula, treated from April 2008 to February 2010. There were 5 patients: 4 suffered gunshot wounds and 1 was stabbed. The mean injury severity score was 22. Initial treatment was insertion of a chest tube in all cases. One patient underwent damage-control surgery and hepatic packing, and 3 were managed with laparotomy, a perihepatic closed drain, and suture of the diaphragm. Two patients developed bronchobiliary fistulas and 3 had pleurobiliary fistulas. Diagnostic procedures involved determination of bilirubin in pleural effusion, computed tomography, magnetic resonance cholangiography, hepatobiliary iminodiacetic scans, and endoscopic retrograde cholangiography. Definitive treatment included sphincterotomy and stenting in 4 cases, pulmonary decortication in 5, fistulectomy in 2, hepatic suture in 2, perihepatic closed drain placement in 4, and suture of the diaphragm in 4. Traumatic thoracobiliary fistulas are complex lesions. A multidisciplinary approach is required for a timely and successful outcome. Endoscopic retrograde cholangiography is very useful as the initial procedure to confirm the diagnosis and also for treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220000
Iminodiacetic acid, 98%
C4H7NO4
56781
Iminodiacetic acid, purum, ≥98.0% (T)
C4H7NO4