EMAIL THIS PAGE TO A FRIEND
International journal of radiation oncology, biology, physics
Detection and repair of ionizing radiation-induced DNA double strand breaks: new developments in nonhomologous end joining.
PMID 23433795
Abstract
DNA damage can occur as a result of endogenous metabolic reactions and replication stress or from exogenous sources such as radiation therapy and chemotherapy. DNA double strand breaks are the most cytotoxic form of DNA damage, and defects in their repair can result in genome instability, a hallmark of cancer. The major pathway for the repair of ionizing radiation-induced DSBs in human cells is nonhomologous end joining. Here we review recent advances on the mechanism of nonhomologous end joining, as well as new findings on its component proteins and regulation.
Related Materials