EMAIL THIS PAGE TO A FRIEND
Experimental Hematology
Isolated Hoxa9 overexpression predisposes to the development of lymphoid but not myeloid leukemia.
PMID 23435313
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-178
Anti-Hox A9 Antibody, clone, Upstate®, from rabbit