EMAIL THIS PAGE TO A FRIEND
European journal of emergency medicine : official journal of the European Society for Emergency Medicine
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
PMID 23435336