PloS one
Protection by salidroside against bone loss via inhibition of oxidative stress and bone-resorbing mediators.
PMID 23437352
Abstract
Oxidative stress is a pivotal pathogenic factor for bone loss in mouse model. Salidroside, a phenylpropanoid glycoside extracted from Rhodiola rosea L, exhibits potent antioxidative effects. In the present study, we used an in vitro oxidative stress model induced by hydrogen peroxide (H(2)O(2)) in MC3T3-E1 cells and a murine ovariectomized (OVX) osteoporosis model to investigate the protective effects of salidroside on bone loss and the related mechanisms. We demonstrated that salidroside caused a significant (P<0.05) elevation of cell survival, alkaline phosphatase (ALP) staining and activity, calcium deposition, and the transcriptional expression of Alp, Col1a1 and Osteocalcin (Ocn) in the presence of H(2)O(2). Moreover, salidroside decreased the production of intracellular reactive oxygen species (ROS), and osteoclast differentiation inducing factors such as receptor activator of nuclear factor-kB ligand (RANKL) and IL-6 induced by H(2)O(2). In vivo studies further demonstrated that salidroside supplementation for 3 months caused a decrease in malondialdehyde (MDA) and an increase in reduced glutathione (GSH) concentration in blood of ovariectomized mouse (P<0.05), it also improved trabecular bone microarchitecture and bone mineral density in the fourth lumbar vertebra and distal femur. Our study indicated that the protection provided by salidroside in alleviating bone loss was mediated, at least in part, via inhibition of the release of bone-resorbing mediators and oxidative damage to bone-forming cells, suggesting that salidroside can be used as an effective remedy in the treatment or prevention of osteoporosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43866
Salidroside, analytical standard
C14H20O7