EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Reply to Ricciotti et al.: Evidence for vascular COX isoforms.
PMID 23437445
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6188
Prostaglandin I2 sodium salt, ≥96% (HPLC), synthetic, powder
C20H31NaO5