EMAIL THIS PAGE TO A FRIEND
Environmental technology
Hydrothermal treatment of incineration fly ash for PCDD/Fs decomposition: the effect of iron addition.
PMID 23437649
Abstract
The catalytic effect of Fe addition on the decomposition of polychlorinated dibenzo-dioxins and polychlorinated dibenzofurans (PCDD/Fs) contained in municipal solid waste incineration (MSWI) fly ash during the hydrothermal process was investigated. Influencing factors, such as Fe addition mode, reaction time and cooling procedure after reaction, were tested to evaluate their effects. Experimental results indicated that Fe addition in the form of a mixture of ferrous sulphate and ferric sulphate enhanced decomposition of PCDD/Fs contained in the MSWI fly ash, particularly for the decomposition of 2,3,7,8-tetrachlorodibenzo-dioxin and 2,3,7,8-tetrachlorodibenzo-furan under the reaction temperature of 563 K. The decomposition rate of PCDD/Fs reached 90.33% by international toxicity equivalent (I-TEQ) when Fe was added as a mixture of ferrous and ferric sulphates by 5% (wt/wt) with the Fe (III)/Fe (II) ratio being 2; without Fe addition, the decomposition rate of PCDD/Fs was only 46.17% by I-TEQ in the same process. Fe addition in the form of ferrous sulphate alone also showed an enhancing effect on PCDD/Fs decomposition, but the associated decomposition rates were relatively lower, suggesting iron oxides formed from the mixture of ferric and ferrous sulphates are more favourable catalysts. At the same time, the cooling procedure after the hydrothermal reaction became more flexible if Fe was added in the form of a mixture of ferric and ferrous sulphates. Although a longer reaction time was helpful to increase decomposition rates of PCDD/Fs, 1 h was proved to be a reasonable time under this condition.