EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Development of three PCR assays for the differentiation between Echinococcus shiquicus, E. granulosus (G1 genotype), and E. multilocularis DNA in the co-endemic region of Qinghai-Tibet plateau, China.
PMID 23438764
Abstract
To investigate echinococcosis in co-endemic regions, three polymerase chain reaction (PCR) assays based on the amplification of a fragment within the NADH dehydrogenase subunit 1 (ND1) mitochondrial gene were optimized for the detection of Echinococcus shiquicus, Echinococcus granulosus G1, and Echinococcus multilocularis DNA derived from parasite tissue or canid fecal samples. Specificity using parasite tissue-derived DNA was found to be 100% except for E. shiquicus primers that faintly detected E. equinus DNA. Sensitivity of the three assays for DNA detection was between 2 and 10 pg. Ethanol precipitation of negative PCR fecal samples was used to eliminate false negatives and served to increase sensitivity as exemplified by an increase in detection from 0% to 89% of E. shiquicus coproDNA using necropsy-positive fox samples.
Related Materials