EMAIL THIS PAGE TO A FRIEND
Journal of occupational and environmental medicine
Association of aryl hydrocarbon receptor gene polymorphism with the neurobehavioral function and autonomic nervous system function changes induced by benzo[a]pyrene exposure in coke oven workers.
PMID 23439269
Abstract
To analyze the association of aryl hydrocarbon receptor (AhR) gene polymorphism and the neurotoxicity induced by benzo[a]pyrene (B[a]P) in coke oven workers. Subjects, 214 coke oven workers and 81 controls, were detected for neurobehavioral function and autonomic nervous system (ANS) function. Airborne B[a]P concentration, urinary 1-hydroxypyrene level, and AhR gene polymorphisms were determined and analyzed for their association with B[a]P neurotoxicity. Neurobehavioral function and ANS function were significantly decreased and dependent on B[a]P dose. The AhR GG, GA, and AA genotypes in G1661A fitted the Hardy-Weinberg equation, whereas C1549T and G1708A gene mutants were not detected. Indices indicating neurotoxicity showed no significant difference among individuals with AA, GG, or GA genotype except for the confusion-bewilderment (P > 0.05). The AhR gene polymorphism is not thought to correlate with B[a]P neurotoxicity among coke oven workers.