EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
High-performance liquid chromatographic separation of acylcarnitines following derivatization with 4'-bromophenacyl trifluoromethanesulfonate.
PMID 2344045
Abstract
A high-performance liquid chromatographic method for the separation of acylcarnitines after derivatization with 4'-bromophenacyl trifluoromethanesulfonate is presented. Derivatization of acylcarnitines was achieved at room temperature within 10 min. Separation of the acylcarnitine 4'-bromophenacyl esters was accomplished by high-performance liquid chromatography using as the analytical column a Resolve-PAK 5-microns C18 radially compressed cartridge eluted with a tertiary gradient containing varying proportions of water, acetonitrile, tetrahydrofuran, triethylamine, potassium phosphate, and phosphoric acid. Acylcarnitine 4'-bromophenacyl esters were detected spectrophotometrically at 254 nm. Baseline separation was obtained for a standard mixture (5 nmol of each injected) containing carnitine, acetyl-, propionyl-, butyryl-, valeryl-, hexanoyl-, heptanoyl-, octanoyl-, nonanoyl-, decanoyl-, lauroyl-, myristroyl-, palmitoyl-, and stearoylcarnitine. Nearly complete separation was obtained for a standard mixture containing butyryl-, isobutyryl-, isovaleryl-, and 2-methylbutyrylcarnitine. The method was applied to a normal human urine and then to this same urine spiked with the acylcarnitine standards. Urinary acylcarnitine profiles from patients having propionic acidemia, isovaleric acidemia, and medium-chain acyl-CoA dehydrogenase deficiency were performed. Urinary isovalerylcarnitine was quantified in the patient with isovaleric acidemia using heptanoylcarnitine as an internal standard.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50405
2-Methylbutyryl-L-carnitine, ≥97.0% (HPLC)
C12H23NO4
51085
Isobutyryl-L-carnitine, ≥97.0% (TLC)
C11H21NO4