EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
In situ synthesis of P3HT-capped CdSe superstructures and their application in solar cells.
PMID 23442609
Abstract
Organic/inorganic hybrid solar cells have great potentials to revolutionize solar cells, but their use has been limited by inefficient electron/hole transfer due to the presence of long aliphatic ligands and unsatisfying continuous interpenetrating networks. To solve this problem, herein, we have developed a one-pot route for in situ synthesis of poly(3-hexylthiophene) (P3HT)-capped CdSe superstructures, in which P3HT acts directly as the ligands. These CdSe superstructures are in fact constructed from numerous CdSe nanoparticles. The presence of P3HT ligands has no obvious adverse effects on the morphologies and phases of CdSe superstructures. Importantly, higher content of P3HT ligands results in stronger photoabsorption and fluorescent intensity of CdSe superstructure samples. Subsequently, P3HT-capped CdSe superstructures prepared with 50 mg P3HT were used as a model material to fabricate the solar cell with a structure of PEDOT:PSS/P3HT-capped CdSe superstructures: P3HT/Al. This cell gives a power conversion efficiency of 1.32%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
752193
1,3,5-Tris(N-carbazolyl)benzene, 97%
C42H27N3