EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
A new hydroxychavicol dimer from the roots of Piper betle.
PMID 23442932
Abstract
A new hydroxychavicol dimer, 2-(g'-hydroxychavicol)-hydroxychavicol (1), was isolated from the roots of Piper betle Linn. along with five known compounds, hydroxychavicol (2), aristololactam A II (3), aristololactam B II (4), piperolactam A (5) and cepharadione A (6). The structures of these isolated compounds were elucidated by spectroscopic methods. Compounds 1 and 2 exhibited inhibitory effects on the generation of superoxide anion and the release of elastase by human neutrophils.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18078
4-Allylpyrocatechol, analytical standard
C9H10O2