EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Tuneable nanochannel formation for sample-in/answer-out devices.
PMID 23443891
Abstract
Control of the dielectric breakdown of PDMS was achieved by limiting the current during the breakdown process. This enabled tuning of the nanochannel pore size and hence their permeability for molecules of different molecular weights. This method enabled the analysis of the drug quinine from whole blood in 3 min using a simple, disposable microfluidic device.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252441
Rhodamine 6G perchlorate, Dye content 99 %
C28H30N2O3 · HClO4