EMAIL THIS PAGE TO A FRIEND
Organic letters
Iodobenzene catalyzed C-H amination of N-substituted amidines using m-chloroperbenzoic acid.
PMID 23444897
Abstract
The oxidative C-H amination of N"-aryl-N'-tosyl/N'-methylsulfonylamidines and N,N'-bis(aryl)amidines has been accomplished using iodobenzene as a catalyst to furnish 1,2-disubstituted benzimidazoles in the presence of mCPBA as a terminal oxidant at room temperature. The reaction is general, and the target products can be obtained in moderate to high yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
273031
3-Chloroperbenzoic acid, ≤77%
C7H5ClO3
I7632
Iodobenzene, 98%
C6H5I