EMAIL THIS PAGE TO A FRIEND
British journal of nursing (Mark Allen Publishing)
Erectile dysfunction following retropubic prostatectomy.
PMID 23448953
Abstract
Prostate cancer is the most common cancer to affect men in the UK. Treatment options depend on the grade of tumour, the patient's co-existing diseases and choice of treatment. One potentially curative option is surgery, specifically a radical retropubic prostatectomy or variation thereof. As a consequence of the surgery, men commonly experience two side-effects: urinary incontinence and erectile dysfunction (ED). This paper outlines the clinical management of ED following surgery and aims to provide an overview of how to assess a man who has developed ED and discuss the various treatment options available, along with the efficacy in terms of recovery of erections.