EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
In vitro biological effects of two anti-diabetic medicinal plants used in Benin as folk medicine.
PMID 23452899
Abstract
Extracts from Polygonum senegalensis (Polygonaceae) and Pseudocedrela kotschyi (Meliaceae) are two important traditionally used medicinal plants in rural Benin to treat many diseases and notably type 2 diabetes. The aim of the study was to investigate the α-glucosidase inhibition, antioxidant and antibacterial activities of those plants extract: Polygonum senegalensis leaves, and Pseudocedrela kotschyi root. Hydro-alcoholic (50%) extracts were analyzed for their phytochemical content and tested for their inhibition potency on α-glucosidase from Saccharomyces cerevisiae. Antioxidant activities were assessed using the DPPH, ORAC, FRAP and DCFH-DA (cell based) assay. Finally, the antibacterial activity was evaluated using MIC determination on four Gram-positive cocci (Bacillus subtilis, Clostridium difficile, Enterococcus faecalis, Staphylococcus aureus), three Gram-negative bacilli (Escherichia coli, Pseudomonas aeruginosa, Klebsiella pneumoniae), and the yeast Candida albicans. Each extract presented significant α-glucosidase inhibition and antioxidant activities. Polygonum senegalensis leaf extracts were the most active in each in vitro assay with an IC50 = 1.5 μg/ml for α-glucosidase inhibition and an IC50 = 6.8 μg/ml for DPPH scavenging, - 4.5 μmol Fe II/g of dry matter - 9366 μmol Trolox / g DW - for FRAP and ORAC values, respectively. IC50 = 2.3 μg GA / ml for DCFH-DA assay. Concerning its antibacterial activity, a growth inhibitory effect was observed only against three Gram negative bacilli: B. subtilis, E. faecalis, S. aureus and the yeast C. albicans at high concentration. The results showed that the semi alcoholic extract of the two studied plants possess α-glucosidase inhibitory activity, antioxidant potency, and low antibacterial effect.