EMAIL THIS PAGE TO A FRIEND
Respiratory physiology & neurobiology
Serotonergic mechanisms are necessary for central respiratory chemoresponsiveness in situ.
PMID 23454177
Abstract
Evidence from in vivo and in vitro experiments conclude that serotonin (5-HT) neurons are involved in and play an important role in central respiratory CO2/H(+) chemosensitivity. This study was designed to assess the importance of 5-HT neurons and 5-HT receptor activation in the frequency and amplitude components of the hypercapnic response of the respiratory network in the unanesthetized perfused in situ juvenile rat brainstem preparation that exhibits patterns of phrenic nerve discharge similar to breathing in vivo. Exposure to a hypercapnic perfusate increased phrenic burst frequency and/or amplitude, the neural correlates of breathing frequency and tidal volume in vivo. Hypercapnic responses were also assessed during exposure to ketanserin (5-HT2 receptor antagonist), and 8-OH-DPAT (inhibiting 5-HT neurons via 5-HT1A autoreceptors). Neither of these drugs substantially altered baseline activity, however, both abolished hypercapnic responses of the respiratory network. These data illustrate that 5-HT neurons and 5-HT receptor activation are not required for respiratory rhythm generation per se, but are critical for CO2 responses in situ, supporting the hypothesis that 5-HT neurons play an important role in central ventilatory chemosensitivity in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H140
(R)-(+)-8-Hydroxy-DPAT hydrobromide, ≥98% (HPLC), solid
C16H25NO · HBr