EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Thermal reaction: the spread of bisphenol S via paper products.
PMID 23454568
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103039
4,4′-Sulfonyldiphenol, 98%
C12H10O4S