EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Alternariol induces abnormal nuclear morphology and cell cycle arrest in murine RAW 264.7 macrophages.
PMID 23454835
Abstract
The mycotoxin alternariol (AOH), a frequent contaminant in fruit and cereal products, is known to induce DNA damage with subsequent cell cycle arrest. Here we elucidated the effects of AOH on stages of cell cycle progression using the RAW 264.7 macrophage model. AOH resulted in an accumulation of cells in the G2/M-phase (4N). Most cells exhibited a large G2 nucleus whereas numbers of true mitotic cells were reduced relative to control. Both cyclin B1 and p-cdc2 levels increased, while cyclin B1 remained in the cytoplasm; suggesting arrest in the G2/M transition point. Remarkably, after exposure to AOH for 24h, most of the cells exhibited abnormally shaped nuclei, as evidenced by partly divided nuclei, nuclear blebs, polyploidy and micronuclei (MN). AOH treatment also induced abnormal Aurora B bridges, suggesting that cytokinesis was interfered within cells undergoing karyokinesis. A minor part of the resultant G1 tetraploid (4N) cells re-entered the S-phase and progressed to 8N cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35758
Alternariol, analytical standard
C14H10O5
A1312
Alternariol from Alternaria sp., ~96%
C14H10O5