EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in medicine
Moving average improves the signal-to-noise ratio of kinetic studies in magnetic resonance spectroscopy.
PMID 2345519
Abstract
Moving average can be used to improve the signal-to-noise ratio of NMR kinetic studies. The method was tested on simulated spectra with time-dependent intensities or peak positions. It was then applied to a series of in vivo spectra, showing the pharmacological effect of heptaminol on the intracellular pH of perfused hearts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000063
Heptaminol hydrochloride, European Pharmacopoeia (EP) Reference Standard
C8H19NO · HCl