EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Structure and biosynthesis of fimsbactins A-F, siderophores from Acinetobacter baumannii and Acinetobacter baylyi.
PMID 23456955
Abstract
Novel chatechol/hydroxamate siderophores (named "fimsbactins") were identified in Acinetobacter baumannii ATCC 17978 and Acinetobacter baylyi ADP1. The major compound, fimsbactin A, was isolated from low-iron cultures of A. baylyi ADP1, and its chemical structure was elucidated by mass spectrometry, and detailed (1)H, (13)C and (15)N NMR spectroscopy. From inverse feeding experiments following HPLC-MS analysis, the structures of five additional derivatives were elucidated. The gene cluster encoding the fimsbactin synthetase (fbs) was identified in both genomes, and mutants in fbs genes in A. baylyi were analyzed, thus allowing prediction of the fimsbactin biosynthesis pathway.