EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Key aromatic residues at subsites +2 and +3 of glycoside hydrolase family 31 α-glucosidase contribute to recognition of long-chain substrates.
PMID 23457711
Abstract
Glycoside hydrolase family 31 α-glucosidases (31AGs) show various specificities for maltooligosaccharides according to chain length. Aspergillus niger α-glucosidase (ANG) is specific for short-chain substrates with the highest k(cat)/K(m) for maltotriose, while sugar beet α-glucosidase (SBG) prefers long-chain substrates and soluble starch. Multiple sequence alignment of 31AGs indicated a high degree of diversity at the long loop (N-loop), which forms one wall of the active pocket. Mutations of Phe236 in the N-loop of SBG (F236A/S) decreased k(cat)/K(m) values for substrates longer than maltose. Providing a phenylalanine residue at a similar position in ANG (T228F) altered the k(cat)/K(m) values for maltooligosaccharides compared with wild-type ANG, i.e., the mutant enzyme showed the highest k(cat)/K(m) value of maltotetraose. Subsite affinity analysis indicated that modification of subsite affinities at +2 and +3 caused alterations of substrate specificity in the mutant enzymes. These results indicated that the aromatic residue in the N-loop contributes to determining the chain-length specificity of 31AGs.