EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Improvement of effectivity of photo disinfection of water from bacterial contaminants in the presence of heterogeneous sensitizers based on phthalocyanines grafted to aminopropyl silicagel].
PMID 23458010
Abstract
The possibility of using 12 heterogeneous sensitizers (HS) based on phthalocyanines covalently grafted to aminopropyl silicagel for disinfection of water from bacteria has been studied. For reliable water quality control the technique of performing bacteriological analysis in the presence of HS beads in the sample has been elaborated. The conditions increasing the efficiency of photo disinfection in the presence of HS were studied. Algorithm for estimation of photo disinfectant effect of HS against bacteria was substantiated. Obtained data confirm the perspective of further studies on the substantiation of the possibility of the application of HS for water disinfection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253103
29H,31H-Phthalocyanine, β-form, 98%
C32H18N8