EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Magnesium orotate elicits acute cardioprotection at reperfusion in isolated and in vivo rat hearts.
PMID 23458194
Abstract
Orotic acid and its salts chronically administered have been shown to significantly improve cardiac function in pathological settings associated with ischemia-reperfusion (I/R) injury. The aim of our study was to investigate the effect of magnesium orotate (Mg-Or) administration at the onset of post-ischemic reperfusion on myocardial function and infarct size (IS). Ex-vivo experiments performed on isolated perfused rat hearts were used to compare Mg-Or administration with a control group (buffer treated), ischemic post-conditioning, orotic acid treatment, and MgCl2 treatment. Mg-Or administration was also investigated in an in-vivo model of regional I/R performed in rats undergoing reversible coronary ligation. The effect of Mg-Or on mitochondrial permeability transition pore (mPTP) opening after I/R was investigated in vitro to gain mechanistic insights. Both ex-vivo and in-vivo experiments showed a beneficial effect from Mg-Or administration at the onset of reperfusion on myocardial function and IS. In-vitro assays showed that Mg-Or significantly delayed mPTP opening after I/R. Our data suggest that Mg-Or administered at the very onset of reperfusion may preserve myocardial function and reduce IS. This beneficial effect may be related to a significant reduction of mPTP opening, a usual trigger of cardiac cell death following I/R.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2750
Orotic acid, ≥98% (titration), anhydrous
C5H4N2O4