EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Sub-acute, moderate-dose, but not short-term, low-dose dietary pre-exposure of mice to perfluorooctanoate aggravates concanavalin A-induced hepatitis.
PMID 23458897
Abstract
Exposure of mice to perfluorooctanoate (PFOA) evokes pronounced hepatomegaly along with significant alterations in both the histological structure and immune status of the liver. The present study was designed to evaluate the effects of this perfluorochemical on immune-mediated liver damage. In this connection, the influence of both sub-acute (10 days), moderate-dose (0.002% w/w=3±0.7mg/kg body weight/day) and short-term (28 days), low-dose (0.00005% w/w=70±2μg/kg body weight/day) dietary pretreatment with PFOA on the development of concanavalin A (Con A)-induced liver damage in mice was examined. With sub-acute, moderate, but not short-term, low-dose exposure, PFOA aggravated the acute liver damage caused by Con A, i.e., elevated serum levels of transaminases and led to more pronounced damage of hepatic tissue. This aggravation was associated with significantly enhanced hepatic level of interleukin-6 (IL-6), but unaltered hepatic levels of tumor necrosis factor-alpha (TNF-α), interferon-gamma (IFN-γ) and interleukin-4 (IL-4). Moreover, hepatic DNA fragmentation was not changed by sub-acute exposure to the moderate-dose. Our findings imply that exposure to PFOA may sensitize hepatic parenchymal cells to other toxicants that activate the hepatic immune system and thereby aggravate liver injury during acute inflammation.