EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
π-Complexes from acyl cyanides and lithium dimethylcuprate(I).
PMID 23459603
Abstract
Rapid injection of pyruvonitrile or benzoyl cyanide into solutions of Me2CuLi in THF-d8 at -100 °C gave complexes that were stable at this temperature. 1D NMR with multiply labelled substrates ((13)C/(15)N) and 2D NMR ((1)H/(13)C) identified them as the first cuprate-carbonyl π-complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269212
Pyruvonitrile, 90%
C3H3NO