EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[State of thiol exchange in patients with psoriasis and its possible correction].
PMID 23461015
Abstract
Experiments on erythrocytes of patients were used to study the influence of remaxol and ademetionin on the exchange of thiols. The values of thioldisulfide exchange indicators were determined in the beginning of the period of observation and at the end of every week within one month. Patients who received hepato-protectors in combination with higher-level thiols showed positive dynamics in disease treatment as compared to patients receiving standard therapy. The concentration of restored glutathione and proteins of hydrosulfide groups and the activity of glutathione peroxidase are informative criteria for estimating the course of disease current and the efficiency of therapy of widespread forms of psoriasis.