EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Removal of airborne toxic chemicals by porous organic polymers containing metal-catecholates.
PMID 23463320
Abstract
Porous organic polymers bearing metal-catecholate groups were evaluated for their ability to remove airborne ammonia, cyanogen chloride, sulphur dioxide, and octane by micro-breakthrough analysis. For ammonia, the metal-catecholate materials showed remarkable uptake under humid conditions.