EMAIL THIS PAGE TO A FRIEND
Future cardiology
Statins for treating stable angina: can statins improve the plaque morphology and angina?
PMID 23463966
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4498
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder
C23H35O7Na · xH2O