EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Total synthesis of 6-deoxyerythronolide B via C-C bond-forming transfer hydrogenation.
PMID 23464668
Abstract
The 14-membered macrolide 6-deoxyerythronolide B is prepared in 14 steps (longest linear sequence) and 20 total steps. Two different methods for alcohol CH-crotylation via transfer hydrogenation are deployed for the first time in target-oriented synthesis. Enyne metathesis is used to form the 14-membered ring. The present approach represents the most concise construction of any erythronolide reported, to date.