EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
Worsening Wenckebach after calcium gluconate injection: not uncommon but frequently missed diagnosis.
PMID 23465875
Abstract
The objective of the study is to demonstrate a common etiology of hyperkalemia and illustrate a potential iatrogenic errors in treatment.