EMAIL THIS PAGE TO A FRIEND
Hospital practice (1995)
Inhibitory effect of apixaban compared with rivaroxaban and dabigatran on thrombin generation assay.
PMID 23466964
Abstract
The effect of the oral direct activated factor X (factor Xa) inhibitor apixaban on tissue factor-induced thrombin generation in human plasma was investigated in vitro using the calibrated automated thrombogram (CAT) method and compared with the oral direct factor Xa inhibitor rivaroxaban and the direct thrombin inhibitor dabigatran. Pooled citrated, anticoagulated, platelet-poor human plasma was spiked with apixaban, rivaroxaban, or dabigatran at concentrations of 0.01 to 10 μM. The inhibitory potencies of the compounds were quantified by 5 CAT parameters: the control thrombin lag time (LT) and time to thrombin peak (TTP) for the doubling of inhibitor concentration (IC2x); and the control endogenous thrombin potential (ETP), thrombin peak, and maximum rate of thrombin generation (Vmax) for the inhibitor concentration, which inhibited 50% (IC50). The inhibitors modified CAT concentration dependently. Their inhibitory potencies, expressed as IC2x LT, IC2x TTP, IC50 ETP, IC50 peak thrombin, and IC50 Vmax, were as follows: 0.10 ± 0.01, 0.19 ± 0.02, 0.65 ± 0.11, 0.089 ± 0.019, and 0.049 ± 0.007 μM for apixaban; 0.049 ± 0.007, 0.070 ± 0.009, 0.43 ± 0.07, 0.048 ± 0.008, and 0.022 ± 0.005 μM for rivaroxaban; and 0.063 ± 0.019, 0.18 ± 0.06, 0.50 ± 0.08, 0.55 ± 0.06, and 0.57 ± 0.27 μM for dabigatran. In summary, apixaban, rivaroxaban, and dabigatran have similar potencies in the prolongation of LT and TTP. The CAT parameters that are related to the rate of thrombin generation during the propagation phase (ie, peak thrombin and Vmax) are more sensitive to activities of apixaban and rivaroxaban than dabigatran. The ETP is the least sensitive parameter for measuring the activities of these inhibitors. Recombinant activated factor VII at 5 and 50 μg/mL reversed the anticoagulant effects of apixaban more at 0.2 μM than at 2 μM. Our study suggests that the CAT method is a sensitive assay to monitor the pharmacodynamic and pharmacokinetic properties of apixaban, rivaroxaban, and dabigatran, and may provide insight into the mechanism of action of these inhibitors. Recombinant activated factor VII may have some potential to reverse the anticoagulant effects of apixaban in vitro.