EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Direct synthesis of pyrroles by dehydrogenative coupling of β-aminoalcohols with secondary alcohols catalyzed by ruthenium pincer complexes.
PMID 23468418