EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Harnessing mutagenic homologous recombination for targeted mutagenesis in vivo by TaGTEAM.
PMID 23470991
Abstract
A major hurdle to evolutionary engineering approaches for multigenic phenotypes is the ability to simultaneously modify multiple genes rapidly and selectively. Here, we describe a method for in vivo-targeted mutagenesis in yeast, targeting glycosylases to embedded arrays for mutagenesis (TaGTEAM). By fusing the yeast 3-methyladenine DNA glycosylase MAG1 to a tetR DNA-binding domain, we are able to elevate mutation rates >800 fold in a specific ∼20-kb region of the genome or on a plasmid that contains an array of tetO sites. A wide spectrum of transitions, transversions and single base deletions are observed. We provide evidence that TaGTEAM generated point mutations occur through error-prone homologous recombination (HR) and depend on resectioning and the error-prone polymerase Pol ζ. We show that HR is error-prone in this context because of DNA damage checkpoint activation and base pair lesions and use this knowledge to shift the primary mutagenic outcome of targeted endonuclease breaks from HR-independent rearrangements to HR-dependent point mutations. The ability to switch repair in this way opens up the possibility of using targeted endonucleases in diverse organisms for in vivo-targeted mutagenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6142 Acc I from Acinetobacter calcoaceticus, Restriction Enzyme
R6885 Alu I from Arthrobacter luteus, Restriction Enzyme
R8631
Bcl I from Bacillus caldolyticus, Restriction Enzyme
R6377
Bgl II from Bacillus licheniformis, Restriction Enzyme
R3131
Bln I from Brevibacterium linens, buffered aqueous glycerol solution
R3635
Bsm I from Bacillus stearothermophilus NUB 36, Restriction Enzyme
R4253 BstE II from Bacillus stearothermophilus ET, Restriction Enzyme
R1761
Cfo I from Clostridium formicoaceticum, Restriction Enzyme
R7763
Cla I from Caryophanon latum L, Restriction Enzyme
R4256
Dde I from Desulfovibrio desulfuricans strain Norway, Restriction Enzyme
R4381
Dra I from Deinococcus radiophilus, Restriction Enzyme
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme
R6265
EcoR I from Escherichia coli BS5, Restriction Enzyme
R2756
EcoR V from Escherichia coli, buffered aqueous glycerol solution
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme
R1632
Mva I from Micrococcus varians Rfl 19, Restriction Enzyme
R8761 Nco I from Nocardia corallina, Restriction Enzyme
R5634
Nhe I from Neisseria mucosa heidelbergensis, Restriction Enzyme
R8506
Not I from Nocardia otidiscaviarum, Restriction Enzyme
R5884 Nsi I from Neisseria sicca, Restriction Enzyme
R7023
Pst I from Providencia stuartii, Restriction Enzyme
R1508
Pvu I from Proteus vulgaris, Restriction Enzyme
R2631
Pvu II from Proteus vulgaris, Restriction Enzyme
R4756 Rsa I from Rhodopseudomonas sphaeroides, Restriction Enzyme
R0754 Sal I from Streptomyces albus G, Restriction Enzyme
R8256
Sfi I from Streptomyces fimbriatus, Restriction Enzyme
R7135 Sph I from Streptomyces phaeochromogenes, Restriction Enzyme
R7260
Xba I from Xanthomonas badrii, Restriction Enzyme
R6379
Xho I from Xanthomonas holcicola, Restriction Enzyme
R1259 Afl III from Anabaena flos-aquae, buffered aqueous glycerol solution