EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Rifaximin is an efficacious treatment for the Parkinsonian phenotype of hepatic encephalopathy.
PMID 23471844
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33999
Rifaximin, VETRANAL, analytical standard
C43H51N3O11
R9904
Rifaximin
C43H51N3O11