EMAIL THIS PAGE TO A FRIEND
Urologic nursing
Adding to the evidence base: application drying time for oxybutynin chloride topical gel.
PMID 23472532
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2881
Oxybutynin chloride, ≥98% (TLC), powder
C22H31NO3 · HCl
O5015
Oxybutynin chloride, meets EP, USP testing specifications
C22H31NO3 · HCl