EMAIL THIS PAGE TO A FRIEND
Current biology : CB
Cytoskeleton: axons earn their stripes.
PMID 23473560
Abstract
Axons must be supported by a strong and flexible cytoskeleton. New 'super-resolution' imaging of the submembranous axonal cytoskeleton reveals that it is organized in a periodic, ladder-like structure with alternating rings of actin linked together by intervening complexes of spectrin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3644 Spectrin from human erythrocytes, buffered aqueous glycerol solution