EMAIL THIS PAGE TO A FRIEND
Postepy higieny i medycyny doswiadczalnej (Online)
[Bacterial type I topoisomerases - biological function and potential use as targets for antibiotic treatments].
PMID 23475489
Abstract
The bacterial chromosome is composed of topologically independent domains, whose spatial organization is controlled by enzymes called topoisomerases. Topology maintenance is crucial in many important cellular processes such as replication, transcription and recombination. Moreover, the role of chromosome topology in adaptation of bacteria to environmental changes and, in the case of pathogenic strains, in their virulence was described. In recent years higher numbers of pathogenic strains resistant to antibiotic treatment have been noticed. In this paper we present the current state of knowledge about the structure and cellular functions of bacterial topoisomerases IA. In particular, we discuss the potential use of these enzymes as new targets for antibacterial compounds.