EMAIL THIS PAGE TO A FRIEND
Journal of Tissue Engineering and Regenerative Medicine
A novel three-dimensional scaffold for regenerative endodontics: materials and biological characterizations.
PMID 23475586