EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[HPLC determination of hydroxysafflor yellow A in Mongolian medicine Dedu Honghuaqiwei pill].
PMID 23477163
Abstract
To establish a method for the determination of hydroxysafflor yellow A in Dedu Honghuaqiwei pill. The determination was performed by HPLC method on Diamonsil C18 (4.6 mm x 250 mm, 5 microm) column at 403 nm using methanol-acetonitrile-0.7% phosphoric acid-water (26: 2: 72) as mobile phase. The column temperature was 30 degrees C and the flow rate was 1.0 mL x min(-1). The linear rang of hydroxysafflor yellow A was 0.068-0.408 microg and the recovery was 97.66%. The result is accurate with good resolution, and the established method can be applied to determine the content of hydroxysafflor yellow A in Dedu Honghuaqiwei pill.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11970
1,3-Diphenyl-2-propenone, ≥98.0% (GC)
C15H12O