Journal of medicinal food
Safety and efficacy of polycalcium for improving biomarkers of bone metabolism: a 4-week open-label clinical study.
PMID 23477624
Polycalcium is a mixture of Polycan and calcium lactate-gluconate 1:9 (w/w) with demonstrated antiosteoporosis activity in vitro and in vivo studies. These studies were a 4-week open-label, single-center trial to evaluate the efficacy of oral Polycalcium on bone metabolism and safety. In total, 30 healthy women (range 40-60 years) were administered 400 mg of Polycalcium for 4 weeks. The primary efficacy parameter was urinary deoxypyridinoline (DPYR) levels, and serum osteocalcin (OSC), bone-specific alkaline phosphatase (BALP), urinary cross-linked C-telopeptide of type-1 collagen (CTx), urinary cross-linked N-telopeptide of type-1 collagen (NTx), calcium (Ca), and phosphorus (P) levels, which were evaluated for comparison before and after administration of Polycalcium. After 4 weeks of Polycalcium administration, 27 subjects completed the test plan. Three subjects withdrew their consent to participate. The values of blood OSC, BALP, serum Ca, and serum P from baseline to 4 weeks of treatment were changed by -28.44%, 14.37%, 6.11%, and 1.42%, respectively. Biomarkers of bone resorption: urinary DPYR, serum CTx, serum NTx, urinary Ca, and urinary P, at baseline after 4 weeks of treatment were changed by -13.40%, 6.67%, -5.13%, -22.43%, and -3.04%, respectively. Additionally, when considering the subjects' adverse effects and the results of the blood and urine tests over the 4-week trial period, the dose of 400  mg Polycalcium showed efficacy for improving bone metabolism and was well tolerated and safe. Polycalcium was apparently safe and efficacious.