EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
5-amino-6-methyluracil is a promising pyrimidine antioxidant.
PMID 23479008