EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Humoral mechanisms regulating the functions of progenitor cells in chronic hepatitis.
PMID 23484186
Abstract
We studied the secretory function of the liver and bone marrow microenvironment cells under conditions of chronic CCl4-induced hepatitis. Enhanced production of substances stimulating parenchymal tissue-specific precursors and stem cell homing factors by liver cells was observed. In situ binding of stem cells by bone marrow elements increased due to production of various cooperation and adhesion factors against the background of reduced SDF-1 secretion.