EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Ultra high performance liquid chromatography/tandem mass spectrometry based identification of steroid esters in serum and plasma: an efficient strategy to detect natural steroids abuse in breeding and racing animals.
PMID 23484650
Abstract
During last decades, the use of natural steroids in racing and food producing animals for doping purposes has been flourishing. The endogenous or exogenous origin of these naturally occurring steroids has since remained a challenge for the different anti-doping laboratories. The administration of these substances to animals is usually made through an intra-muscular pathway with the steroid under its ester form for a higher bioavailability and a longer lasting effect. Detecting these steroid esters would provide an unequivocal proof of an exogenous administration of the considered naturally occurring steroids. A quick analytical method able to detect at trace level (below 50 pg/mL) a large panel of more than 20 steroid esters in serum and plasma potentially used for doping purposes in bovine and equine has been developed. Following a pre-treatment step, the sample is submitted to a solid phase extraction (SPE) before analysis with UPLC-MS/MS. The analytical method's efficiency has been probed through three different in vivo experiments involving testosterone propionate intra-muscular administration to three heifers, 17-estradiol benzoate intra-muscular administration to a bull and a heifer and nandrolone laurate intra-muscular administration to a stallion. The results enabled detecting the injected testosterone propionate and 17-estradiol benzoate 2 and 17 days, respectively, post-administration in bovine and nandrolone laurate up to 14 days post-administration in equine. The corresponding elimination profiles in bovine serum and equine plasma have been established. The first bovine experiment exhibited a maximal testosterone propionate concentration of 400 pg/mL in one of the three heifer serum within 5h post-administration. The second bovine experiment reported a maximal 17-estradiol benzoate concentration of 480 pg/mL in the same matrix recorded 9 days after its administration. The last equine experiment resulted in a maximal nandrolone laurate concentration of 440 pg/mL in horse plasma 24h after administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34851
Acetonitrile, for HPLC, gradient grade, ≥99.9%
C2H3N
34998
Acetonitrile, HPLC Plus, ≥99.9%
C2H3N
439134
Acetonitrile, for HPLC, gradient grade, ≥99.9%
C2H3N
360457
Acetonitrile, ACS reagent, ≥99.5%
C2H3N
494445
Acetonitrile, biotech. grade, ≥99.93%
C2H3N
34881
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
C2H3N
33019
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
C2H3N
34442
Acetonitrile, for synthesis of DNA, ≥99.9% (GC)
C2H3N
110086
Acetonitrile, ReagentPlus®, 99%
C2H3N
565423
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
319902
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
34858
Ethyl acetate, for HPLC, ≥99.7%
C4H8O2
650528
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C4H8O2
439169
Ethyl acetate, for HPLC, ≥99.8%
C4H8O2
33211
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
27227
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
C4H8O2
676810
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
494518
Ethyl acetate, biotech. grade, ≥99.8%
C4H8O2
45767
Ethyl acetate, puriss. p.a., free of higher boiling impurities, ≥99.9% (GC)
C4H8O2
47947 Ethyl acetate solution, certified reference material, 2000 μg/mL in methanol
T0300000
Testosterone propionate, European Pharmacopoeia (EP) Reference Standard
C22H32O3