EMAIL THIS PAGE TO A FRIEND
Organic letters
Catalytic enantioselective desymmetrization of meso cyclic anhydrides via iridium-catalyzed hydrogenation.
PMID 23485036
Abstract
A novel method to desymmetrize meso-anhydrides into lactones via asymmetric hydrogenation catalyzed by the Ir-C3*-TunePhos complex has been developed. Various chiral lactones were synthesized with full conversion and excellent enantioselectivity under high reaction temperature.