EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
Rapid and efficient generation of neurons from human pluripotent stem cells in a multititre plate format.
PMID 23486189
Abstract
Existing protocols for the generation of neurons from human pluripotent stem cells (hPSCs) are often tedious in that they are multistep procedures involving the isolation and expansion of neural precursor cells, prior to terminal differentiation. In comparison to these time-consuming approaches, we have recently found that combined inhibition of three signaling pathways, TGFβ/SMAD2, BMP/SMAD1, and FGF/ERK, promotes rapid induction of neuroectoderm from hPSCs, followed by immediate differentiation into functional neurons. Here, we have adapted our procedure to a novel multititre plate format, to further enhance its reproducibility and to make it compatible with mid-throughput applications. It comprises four days of neuroectoderm formation in floating spheres (embryoid bodies), followed by a further four days of differentiation into neurons under adherent conditions. Most cells obtained with this protocol appear to be bipolar sensory neurons. Moreover, the procedure is highly efficient, does not require particular expert skills, and is based on a simple chemically defined medium with cost-efficient small molecules. Due to these features, the procedure may serve as a useful platform for further functional investigation as well as for cell-based screening approaches requiring human sensory neurons or neurons of any type.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10852
Mowiol® 10-98, Mw ~61,000
81365
Mowiol® 18-88, Mw ~130,000
11773
Mowiol® 20-98, Mw ~125,000
10849
Mowiol® 28-99, Mw ~145,000
81381
Mowiol® 4-88, Mw ~31,000
81382
Mowiol® 4-98, Mw ~27,000
324590
Mowiol® 40-88, average Mw ~205,000 g/mol
10851
Mowiol® 56-98, Mw ~195,000
10853
Mowiol® 6-98, Mw ~47,000
81383
Mowiol® 8-88, Mw ~67,000
341584
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
363146
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
563900
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
363065
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
363138
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
363154
Poly(vinyl alcohol), average Mw 85,000-124,000, 98-99% hydrolyzed
363162
Poly(vinyl alcohol), average Mw 146,000-186,000, 98-99% hydrolyzed
348406
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
363111
Poly(vinyl alcohol), average Mw 85,000-124,000, 96% hydrolyzed
363170
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
363073
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
363081
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
363103
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
360627
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
P8136
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
P1763
Poly(vinyl alcohol), Fully hydrolyzed