EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Reactions of organolithiums with dialkyl oxalates. A flow microreactor approach to synthesis of functionalized α-keto esters.
PMID 23486774
Abstract
Reactions of functionalized aryllithiums with dialkyl oxalates were achieved using a flow microreactor to obtain α-keto esters with high selectivity by virtue of fast 1 : 1 micromixing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B57702
Bromobenzene, ReagentPlus®, 99%
C6H5Br
16350
Bromobenzene, ≥99.5% (GC)
C6H5Br